Order Entry
Spain
Orders LinkContactUsLinkComponent
Reference standards VOA

Reference standards VOA

Ethylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
Catalog # REST30290
Supplier:   Restek
Reference standards VOA
Catalog # REST30290
Supplier:  Restek
CAS Number:  
Reference standards VOA

Ethylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL

CAS Number:  
Price
53,00 €
Log in for availability
1 * 1 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Ethylbenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
  • Cat. No.:
    REST30290
  • Formula:
    C₈H₁₀
  • Description:
    Ethylbenzene
  • CAS No.:
    100-41-4

Specifications