Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Reference standards VOA

1,1-dichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
  REST30278
 :   Restek
Reference standards VOA
  REST30278
 :  Restek
 :  
Reference standards VOA

1,1-dichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL

 :  
 
54,50 €
   
1 * 1 mL
 
 

 

  • Pk:
    1 mL
  • Description:
    1,1-dichloroethene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
  • Cat. No.:
    REST30278
  • Formula:
    C₂H₂Cl₂
  • Description:
    1,1-dichloroethene
  • CAS No.:
    75-35-4