Reference standards VOA
4-chlorotoluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30269
Jump to:
Discontinued
Product Details & Documents
Specifications
Specifications
- Env:1
- Descripción:4-chlorotoluene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30269
- Formula:C₇H₇Cl
- Descripción:4-chlorotoluene
- CAS No.:106-43-4



