Order Entry
Spain
Orders LinkContactUsLinkComponent
Reference standards VOA

Reference standards VOA

Chloroform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30266
Supplier:   Restek
Reference standards VOA
Catalog # REST30266
Supplier:  Restek
CAS Number:  
Reference standards VOA

Chloroform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1

CAS Number:  
Price
53,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Description:
    Chloroform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
  • Cat. No.:
    REST30266
  • Formula:
    CHCl₃
  • Description:
    Chloroform
  • CAS No.:
    67-66-3

Specifications