Reference standards VOA
Acetone, 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
Catálogo # REST30245
Proveedor: Restek
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 mL
- Description:Acetone, 5000 µg/ml, purge and trap grade methanol, 1 ml/ampule
- Cat. No.:REST30245
- Formula:C₃H₆₀
- Description:Acetone
- CAS No.:67-64-1
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



