Order Entry
Spain
Orders LinkContactUsLinkComponent
Reference standards VOA

Reference standards VOA

Fluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
Catalog # REST30030
Supplier:   Restek
Reference standards VOA
Catalog # REST30030
Supplier:  Restek
CAS Number:  
Reference standards VOA

Fluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL

CAS Number:  
Price
52,50 €
Log in for availability
1 * 1 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Fluorobenzene, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule

Specifications