Order Entry
Spain
Orders LinkContactUsLinkComponent
Reference standards VOA

Reference standards VOA

Ethylbenzene-d10, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL
Catalog # REST30029
Supplier:   Restek
Reference standards VOA
Catalog # REST30029
Supplier:  Restek
CAS Number:  
Reference standards VOA

Ethylbenzene-d10, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1 mL

CAS Number:  
Price
54,50 €
Log in for availability
1 * 1 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Ethylbenzene-d10, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.