γ-Cyclodextrine
Glass bottle for solids, 5 g
Supplier: MP Biomedicals
Jump to:
Product Details & Documents
Synonyms: gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
- CAS Number:17465-86-0
- Formula:C₄₈H₈₀O₄₀
- MDL Number:MFCD00009595
- Molecular Weight:1297.1248 g/mol
- EINECS:241-482-4
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



