O-RING 45MM x 3MM 1 * 1 items
Catalog # VACU23120544
Supplier: VACUUBRAND
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: Brand
VWR® Reagent Reservoirs
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Accessories for Transferpette® electronic pipettes
Supplier: Brand
O-RINGS FOR NOSE CONE 1 * 24 items
Supplier: EPPENDORF
Accessories for accu-jet® pro
Supplier: Brand
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
Accessories for water purification millipore
Supplier: Merck
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for CO2 Incubators, Heracell VIOS
Supplier: Thermo Scientific