Order Entry
Spain
Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
Electrode cable, 2×B, with banana plug (Ø 4 mm), for connecting electrodes with Metrohm plug-in head G to measuring instrument, 1 m
Catalog # METR6.2104.080
Supplier:   METROHM
Electrode Cables
Catalog # METR6.2104.080
Supplier:  METROHM
Electrode Cables
Electrode cable, 2×B, with banana plug (Ø 4 mm), for connecting electrodes with Metrohm plug-in head G to measuring instrument, 1 m
Price
Inquire For Price
Log in for availability
1 * 1 UN
Product Details & Documents
Specifications
  • Description:
    Electrode cable, 2×B, with banana plug (Ø 4 mm), for connecting electrodes with Metrohm plug-in head G to measuring instrument
  • Length:
    1 m
  • Seq:
    10