Order Entry
Spain
ContactUsLinkComponent
SONOREX LONGLIFE RK 512 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 280X234X200 MM 13,1 L, OSCILLATING TANK OF 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 1 * 1 items
Catalog # BAND3034
SONOREX LONGLIFE RK 512 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 280X234X200 MM 13,1 L, OSCILLATING TANK OF 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 1200 1 * 1 items
Catalog # BAND3034
Frequently Bought Together