Order Entry
Spain
ContactUsLinkComponent
SONOREX LONGLIFE RK 255 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 280X150X150 MM 6,3 L, OSCILLATING TANK OF 2 MM S/S AISI 3 16 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 720 W 1 * 1 items
Catalog # BAND3033
SONOREX LONGLIFE RK 255 CH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 280X150X150 MM 6,3 L, OSCILLATING TANK OF 2 MM S/S AISI 3 16 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C (86-176F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 720 W 1 * 1 items
Catalog # BAND3033
Frequently Bought Together