Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Formaldehyde 4%

Plastic bottle, 10 L
Catalog # PRLSMC313613.9010
Health
Irritant
Supplier:   PROLABO CMP
Formaldehyde 4%
Catalog # PRLSMC313613.9010
Supplier:  PROLABO CMP
CAS Number:  50-00-0
Formaldehyde 4%

Plastic bottle, 10 L

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents

(10% Formaline solution)

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 L

Specifications