Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Formaldehyde 4%

10 L
Catalog # PRLSMC313613.6010
Health
Irritant
Supplier:   PROLABO CMP
Formaldehyde 4%
Catalog # PRLSMC313613.6010
Supplier:  PROLABO CMP
CAS Number:  50-00-0
Formaldehyde 4%

10 L

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents

(10% Formaline solution)

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specifications

  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.