Order Entry
Spain
Orders LinkContactUsLinkComponent
Trolley, trays

Trolley, trays

Trays trolley, without guard rails, two handles and three trays, argon welded, 430W×600L mm, 3
Catalog # AGEN446S
Supplier:   Agencinox
Trolley, trays
Catalog # AGEN446S
Supplier:  Agencinox
Trolley, trays
Trolley, trays

Trays trolley, without guard rails, two handles and three trays, argon welded, 430W×600L mm, 3

Price
1 200,00 €
Log in for availability
1 * 1 UN
Product Details & Documents

Trolleys with two handles, shaped trays made up of 18/10 stainless steel with bright polish finishing.

  • Heavy duty structure
  • Easy cleaning
  • Available in knockdown construction or argon welded models
Specifications
  • Description:
    Trays trolley, without guard rails, two handles and three trays, argon welded, 430W×600L mm
  • Seq:
    06
  • Shelves:
    3
Recomendaciones

Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".

De lo contrario, recibirá recomendaciones genéricas.