Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Test tubes without rim, heavy wall

140 mm, 14 mm, 1,0 mm
  212-7448
Test tubes without rim, heavy wall
  212-7448
 :  
Test tubes without rim, heavy wall
Test tubes without rim, heavy wall

140 mm, 14 mm, 1,0 mm

 

PYREX®, borosilicate glass.

  • Excellent chemical resistance
  • High temperature resistance

High quality test tubes designed and constructed for optimum performance. Medium walled, robust.

ISO 4142

 : ISO 4142

 
  • Ext.Ø:
    14 mm
  • Wall thickness:
    1,0 mm
  • Length:
    140 mm
  • Seq:
    08