Order Entry
Spain
Orders LinkContactUsLinkComponent
Trolley, trays

Trolley, trays

Trays trolley, with guard rails, two handles and two trays, knockdown construction, 500W×750L mm, 2
Catalog # 139-0102
Supplier:   Agencinox
Trolley, trays
Catalog # 139-0102
Supplier:  Agencinox
Trolley, trays
Trolley, trays

Trays trolley, with guard rails, two handles and two trays, knockdown construction, 500W×750L mm, 2

Price
753,00 €
Log in for availability
1 * 1 UN
Product Details & Documents

Trolleys with two handles, shaped trays made up of 18/10 stainless steel with bright polish finishing.

  • Heavy duty structure
  • Easy cleaning
  • Available in knockdown construction or argon welded models
Specifications
  • Description:
    Trays trolley, with guard rails, two handles and two trays, knockdown construction, 500W×750L mm
  • Seq:
    04
  • Shelves:
    2
Recomendaciones

Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".

De lo contrario, recibirá recomendaciones genéricas.