Clamps
60 mm
Catalog # 241-1112
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel.
- Jaws are covered with a layer of cork to protect clamped items
- Without bosshead for fixing to stand
- Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894
Specifications
- Spanning width:60 mm