Order Entry
Spain
Orders LinkContactUsLinkComponent
Papain

Papain

5 kg
Catalog # 1.07149.5000
Papain
Catalog # 1.07149.5000
CAS Number:  
CAS Number:  
Price
2 710,00 €
Log in for availability
1 * 5 kg
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    5 kg
  • Cat. No.:
    1.07149.5000
  • Enzyme activity:
    30000 USP-U/mg
  • Tested applications:
    Suitable for use as excipient
  • Enzyme name:
    Papain

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.