Order Entry
Spain
ContactUsLinkComponent
Eichöl (mineral) B11000, 11000 mPa.s, 1 * 475 ml
Catalog # 616-3132
Supplier:  AMETEK
Eichöl (mineral) B11000, 11000 mPa.s, 1 * 475 ml
Catalog # 616-3132
Supplier:  AMETEK
Frequently Bought Together