Order Entry
Spain
Orders LinkContactUsLinkComponent
Burners
Burners
Teclu burner with stopcock, Propane
  451-1350
Burners
  451-1350
Burners
Burners
Teclu burner with stopcock, Propane
73,90 €
1 * 1 UN
DIN type.
  • Air regulation and pilot flame inside
Ordering information: Supplied without safety base, this needs to be ordered separately.
  • Description:
    Teclu burner with stopcock
  • For:
    Propane