Order Entry
Spain
Orders LinkContactUsLinkComponent
Buffer Solutions

Buffer Solutions

6,00, ±0.01, 25 °C, 4 L
Catalog # 42418.K7
Buffer Solutions
Catalog # 42418.K7
CAS Number:  
Buffer Solutions
Buffer Solutions

6,00, ±0.01, 25 °C, 4 L

CAS Number:  
Price
93,50 €
Log in for availability
1 * 4 L
Specifications

Specifications

  • Pk:
    4 L
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    6,00
  • Cat. No.:
    42418.K7
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00134130

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.