Order Entry
Spain
Orders LinkContactUsLinkComponent
Fused quartz microscope slides
Fused quartz microscope slides
Microscope slides, Fused quartz, 26×51 mm
Catálogo # 42296.KG
Fused quartz microscope slides
Catálogo # 42296.KG
Fused quartz microscope slides
Microscope slides, Fused quartz, 26×51 mm
Price
69,50 €
Log in for availability
1 * 1 UN
Product Details & Documents
These fused quartz slides are made of fusing naturally occurring crystalline silica.
  • High chemical purity, high corrosion resistance, high melting point
  • Low coefficient of thermal expansion with excellent electrical insulation
  • Optical transmission from ultra-violet to infrared
Specifications
  • Description:
    Microscope slides
  • L×W:
    26×51 mm
  • Material:
    Fused quartz
  • Seq:
    02