Order Entry
Spain
Orders LinkContactUsLinkComponent
Flat seals
Flat seals
Flat seal, fits outer thread, 19,05 mm, PE
Catalog # 223-1142
Supplier:   BURKLE
Flat seals
Catalog # 223-1142
Supplier:  BURKLE
Flat seals
Flat seals
Flat seal, fits outer thread, 19,05 mm, PE
Price
4,70 €
Log in for availability
1 * 10 UN
Product Details & Documents
These flat seals are made from transparent PE.
  • Fits outer or inner thread
Specifications
  • Description:
    Flat seal, fits outer thread, 19,05 mm
  • Material:
    PE