Affinity chromatography columns, HiTrap™ protein G HP
HiTrap™ G HP, 34 µm, 16×25 mm, 5 ml, 20 ml/min
Supplier: Cytiva
Jump to:
Product Details & Documents
HiTrap™ Protein G HP affinity columns are pre-packed with 1 or 5 ml of Protein G Sepharose™ high performance for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants.
- Can be operated with a syringe, peristaltic pump or liquid chromatography system such as ÄKTAdesign™ or FPLC™ system
- Polypropylene column material which is biocompatible and non interactive with biomolecules
- Top and bottom frits are manufactured from porous polyethylene
Delivery information: Delivered with a stopper on the inlet and a snap-off end on the outlet.
Documents
Certificates
Search by Lot Number17-0404-01_42 BR 17-0404-01_42 BR (1) 17-0404-01_42 BR (2) 17-0404-01_42 DF 17-0404-01_42 HB
Datasheet
More
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Flow rate:20 ml/min
- Particle Size:34 µm
- Capacity:5 ml
- Description:HiTrap™ G HP
- Seq:04
- Bed dimensions:16×25 mm
- Cat. No.:17-0405-01
- Max. pressure:5 bar
- Storage:2 to 8 °C, 20% Ethanol
- pH range:3 - 9



