Order Entry
Spain
Orders LinkContactUsLinkComponent
Probes for 845

Probes for 845

TC K-type flexible for immersion probe, −200…+1000 °C, ±1.5 °C, 1,5×500 mm
Catalog # 620-3426
Supplier:   Testo
Probes for 845
Catalog # 620-3426
Supplier:  Testo
Probes for 845
Probes for 845

TC K-type flexible for immersion probe, −200…+1000 °C, ±1.5 °C, 1,5×500 mm

Price
34,00 €
Log in for availability
1 * 1 UN
Specifications
  • Range:
    −200…+1000 °C
  • Description:
    TC K-type flexible for immersion probe
  • Seq:
    26
  • Accuracy:
    ±1.5 °C
  • Ø×L:
    1,5×500 mm