Order Entry
Spain
Orders LinkContactUsLinkComponent
Probes for 845

Probes for 845

Probe, magnetic probe for measurement on metal surfaces, TC type K, −50...+170 °C, Class 2 (EN 60584-2), 35 mm, 20×35 mm
  628-3216
 :   Testo
Probes for 845
  628-3216
 :  Testo
Probes for 845
Probes for 845

Probe, magnetic probe for measurement on metal surfaces, TC type K, −50...+170 °C, Class 2 (EN 60584-2), 35 mm, 20×35 mm

 
181,00 €
   
1 * 1 UN
 
  • Range:
    −50...+170 °C
  • Description:
    Probe, magnetic probe for measurement on metal surfaces, TC type K
  • Length:
    35 mm
  • Seq:
    11
  • Accuracy:
    Class 2 (EN 60584-2)
  • Application:
    Surface
  • Ø×L:
    20×35 mm