Order Entry
Spain
Orders LinkContactUsLinkComponent
Probes for 845

Probes for 845

Fibreglass flexible thermocouple with TC adapter, −50…+400 °C, −40...+1200 °C, 1,5×1500 mm
Catalog # 628-3212
Supplier:   Testo
Probes for 845
Catalog # 628-3212
Supplier:  Testo
Probes for 845
Probes for 845

Fibreglass flexible thermocouple with TC adapter, −50…+400 °C, −40...+1200 °C, 1,5×1500 mm

Price
34,00 €
Log in for availability
1 * 1 UN
Specifications
  • Range:
    −50…+400 °C
  • Description:
    Fibreglass flexible thermocouple with TC adapter
  • Seq:
    24
  • Accuracy:
    −40...+1200 °C
  • Application:
    Surface
  • Ø×L:
    1,5×1500 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.