Order Entry
Spain
Orders LinkContactUsLinkComponent
L(+)-Glutamic acid, Millipore®

L(+)-Glutamic acid, Millipore®

Plastic drum, 10 kg
Catalog # 1.00291.9010
Supplier:   Merck
L(+)-Glutamic acid, Millipore®
Catalog # 1.00291.9010
Supplier:  Merck
CAS Number:  56-86-0
L(+)-Glutamic acid, Millipore®
L(+)-Glutamic acid, Millipore®

Plastic drum, 10 kg

CAS Number:  56-86-0
Price
2 470,00 €
Log in for availability
1 * 10 kg
Product Details & Documents

Soluble in water and mineral acids; insoluble in organic solvents. In aqueous solutions forms pyrrolidonecarboxylic acid slowly at room temperature, more rapidly at 100 °C.

Synonyms:  (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
  • CAS Number:56-86-0
  • Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00002634
  • Molecular Weight:147.13 g/mol
  • Merck Index:13,04482
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:160 °C
  • Density:1.538 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-293-7
Specifications

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 kg

Specifications