Tannic acid 95%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Gallotannin, Tannin
- CAS Number:1401-55-4
- Formula:C₇₆H₅₂O₄₆
- MDL Number:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Melting Pt:218 °C (dec.)
- Density:Min, 1 g/cm³ (20 °C)
- Flash Pt:198 °C
- Storage Temperature:Ambient
- EINECS:215-753-2
Documents
Specifications
Specification Test Results
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- Assay:≥94 % (on dry substance)
- Loss on drying:≤12.0 %
- Sulfated ash:≤1.0 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



