BD Discardit™ II Syringes
Syringe, Luer slip, eccentric tip, 20 ml, Sterile
The BD Discardit™ II two-piece syringe is a single-use medical device intended for injection and aspiration of medical fluids, including both bodily fluids (blood, etc.) and medicines. The lubricant included in the barrel material allows smooth advancement of the plunger, leading to reduced sliding force.
- Clear and transparent barrel for good visualisation of syringe content
- Leak-tight
- Accurate graduation suited to user's needs
- Robust retaining ring to prevent accidental plunger rod pullout during aspiration
- Design of finger grip flanges ensures stability and comfort during injection
- Ergonomically designed plunger for single-handed operation
- Sterile, single-use
- Natural rubber latex is not part of the material formulation
- Sterility:Sterile
- Capacity:20 ml
- Description:Syringe, Luer slip, eccentric tip
- Seq:10