Strychnine 98%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:57-24-9
- Formula:C₂₁H₂₂N₂O₂
- MDL Number:MFCD00005941
- Molecular Weight:334.42 g/mol
- Melting Pt:284…286 °C
- Storage Temperature:Ambient
- EINECS:200-319-7
- UN:1692
- ADR:6.1,I
Documents
ACRO15895
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
- Chloride (Cl):≤0.01 %
- Iron (Fe):≤0.001 %
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Glass bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



