Thymolphthalein, pure indicator
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Thymolphthalein, pure indicator
Catalog # ACRO151460100
Supplier: Thermo Fisher Scientific
CAS Number: 125-20-2
Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
- CAS Number:125-20-2
- Formula:C₂₈H₃₀O₄
- MDL Number:MFCD00005909
- Molecular Weight:430.54 g/mol
- Merck Index:12,09542
- Melting Pt:251…253 °C
- Storage Temperature:Ambient
- EINECS:204-729-7
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- Sulfated ash:≤0.5 %
- Visual transition interval:from pH 8.8 (colorless) to pH 10.5 (blue)
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



