Order Entry
Spain
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA

3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA

Glass bottle, 100 mL
Catalog # ACRO328051000
3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA
Catalog # ACRO328051000
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA
3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA

Glass bottle, 100 mL

CAS Number:  54827-17-7
Price
262,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow-green to light blue Solution
  • pH:
    3.50 to 3.90
  • Activity:
    90 to 110 %
  • Absorbance:
    ≤0.025 (450/620 nm, 0.2M H2SO4)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.