Order Entry
Spain
Orders LinkContactUsLinkComponent
 

3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate

Glass bottle, 100 mL
Catalog # ACRO328031000
3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate
Catalog # ACRO328031000
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate
3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate

Glass bottle, 100 mL

CAS Number:  54827-17-7
Price
144,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow-green to light blue Solution
  • Absorbance:
    ≤0.05 (450/620 nm, 0.2M H2SO4)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications