Order Entry
Spain
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate

3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate

Glass bottle, 30 mL
Catalog # ACRO328030300
3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate
Catalog # ACRO328030300
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate
3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate

Glass bottle, 30 mL

CAS Number:  54827-17-7
Price
54,80 €
Log in for availability
1 * 30 mL
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow-green to light blue Solution
  • Absorbance:
    ≤0.05 (450/620 nm, 0.2M H2SO4)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    30 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.