Order Entry
Spain
Orders LinkContactUsLinkComponent
Clamps
Clamps
120
Catalog # 241-0402
Supplier:   BOCHEM
Clamps
Catalog # 241-0402
Supplier:  BOCHEM
Clamps
Price
66,40 €
Log in for availability
1 * 1 UN
Product Details & Documents
Stainless steel.
  • Jaws are covered with a layer of cork to protect clamped items
  • Without bosshead for fixing to stand
  • Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894
Specifications
  • Spanning width:
    120