Lead 99+%, granules
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7439-92-1
- Formula:Pb
- MDL Number:MFCD00134050
- Molecular Weight:207.2 g/mol
- Merck Index:12,05410
- Boiling Pt:1740 °C (1013 hPa)
- Melting Pt:327.4 °C
- Density:11,3437 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-100-4
Documents
ACRO22262
SDS
Specifications
Specification Test Results
- Appearance:Grey Granules
- Assay:≥99.0 %
Specifications
- Pack type:Plastic bottle
- Pk:100 g



