Dressing forceps, standard
Standard, Straight, blunt, 160 mm
Catalog # HAMML-HSC056-16
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Stainless steel.
Specifications
- Type:Straight, blunt
- Description:Standard
- Length:160 mm
- Seq:05