Order Entry
Spain
Orders LinkContactUsLinkComponent
Anion exchange bulk packing material

Anion exchange bulk packing material

PRP-X600 bulk resin, 7 µm, -, 1 g
Catalog # 553-0018
Supplier:   HAMILTON BONADUZ
Anion exchange bulk packing material
Catalog # 553-0018
Supplier:  HAMILTON BONADUZ
CAS Number:  
Anion exchange bulk packing material
Anion exchange bulk packing material

PRP-X600 bulk resin, 7 µm, -, 1 g

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents

Polymer bulk packing material for anion exchange applications.

Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.

Specifications

Specifications

  • Particle Size:
    7 µm
  • Pk:
    1 g
  • Pore size:
    -
  • Description:
    PRP-X600 bulk resin

Specifications