Entrada de pedido
Spain
Orders LinkContactUsLinkComponent
 

Anion exchange bulk packing material

PRP-X100 bulk resin, 12 - 20 µm, 100 Å, 1 g
Catalog # 553-0007
Supplier:   HAMILTON BONADUZ
Anion exchange bulk packing material
Catalog # 553-0007
Supplier:  HAMILTON BONADUZ
CAS Number:  
Anion exchange bulk packing material
Anion exchange bulk packing material

PRP-X100 bulk resin, 12 - 20 µm, 100 Å, 1 g

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents

Polymer bulk packing material for anion exchange applications.

Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.

Specifications

Specifications

  • Tamaño de partícula:
    12 - 20 µm
  • Env:
    1 g
  • Tamaño de poro:
    100 Å
  • Descripción:
    PRP-X100 bulk resin

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.