Sulfate standard solution
Sulfate standard solution, 500 ml (NIST), 1000 mg/l as SO42-, 500 mL
: Hach
Standard solution for accuracy checks in sulfate determinations.
- Concentration:1000 mg/l as SO<sub>4</sub><sup>2-</sup>
- Pk:500 mL
- Description:Sulfate standard solution, 500 ml (NIST)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



