5415D Spare Part - Cable Braking resistor 230V includes resistor 150R/50W and connecting cable - 1/pack 1 * 1 items
EPPE5425601.000
5415D Spare Part - Cable Braking resistor 230V includes resistor 150R/50W and connecting cable - 1/pack 1 * 1 items
EPPE5425601.000
Frequently Bought Together
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection