Order Entry
Spain
Orders LinkContactUsLinkComponent
Grape Seed Extract

Grape Seed Extract

Bottle
Catalog # APOSBUP16617-5G
New Product
Supplier:   Apollo Scientific
Grape Seed Extract
Catalog # APOSBUP16617-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  84929-27-1
Grape Seed Extract

Bottle

CAS Number:  84929-27-1
Price
414,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Grape Seed P.E.
  • CAS Number:84929-27-1
  • Formula:C₃₂H₃₀O₁₁
  • Molecular Weight:590,581 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.