Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Grape Seed Extract 95%

Bottle
Catalog # APOSOR1028071-100G
New Product
Supplier:   Apollo Scientific
Grape Seed Extract 95%
Catalog # APOSOR1028071-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  84929-27-1
Grape Seed Extract 95%

Bottle

CAS Number:  84929-27-1
Price
72,40 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Grape Seed P.E.
  • CAS Number:84929-27-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications