Order Entry
Spain
Orders LinkContactUsLinkComponent
Grape Seed Extract 95%

Grape Seed Extract 95%

Bottle
Catalog # APOSOR1028071-5G
New Product
Supplier:   Apollo Scientific
Grape Seed Extract 95%
Catalog # APOSOR1028071-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  84929-27-1
Grape Seed Extract 95%

Bottle

CAS Number:  84929-27-1
Price
36,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Grape Seed P.E.
  • CAS Number:84929-27-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.