Cesium chloride
Bottle
Catalog # APOSBUP19587-1G
New Product
Supplier: Apollo Scientific
Cesium chloride
Catalog # APOSBUP19587-1G
Supplier: Apollo Scientific
New Product
CAS Number: 7647-17-8
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: Cesium Chloride
- CAS Number:7647-17-8
- Formula:ClCs
- Molecular Weight:168,36 g/mol
Specifications
Specifications
- Pack type:Bottle



