Order Entry
Spain
Orders LinkContactUsLinkComponent
MOPS ≥98%

MOPS ≥98%

Bottle
Catalog # APOSBUP18762-500MG
New Product
Supplier:   Apollo Scientific
MOPS ≥98%
Catalog # APOSBUP18762-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1132-61-2
MOPS ≥98%

Bottle

CAS Number:  1132-61-2
Price
37,70 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
  • CAS Number:1132-61-2
  • Formula:C₇H₁₅NO₄S
  • Molecular Weight:209,26 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.