Order Entry
Spain
Orders LinkContactUsLinkComponent
Grape Seed Extract

Grape Seed Extract

New Product
Supplier:   Apollo Scientific
Grape Seed Extract
Supplier:  Apollo Scientific
New Product
Certificates
Grape Seed Extract

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Grape Seed P.E.
  • CAS Number:84929-27-1
  • Formula:C₃₂H₃₀O₁₁
  • Molecular Weight:590,581 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 57,90 €
    57,90 €
    1 * 100 mg
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 414,00 €
    414,00 €
    1 * 5 g
       414,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.