Order Entry
Spain
Orders LinkContactUsLinkComponent
Blocks for PCR
Blocks for PCR
New Product
Supplier:   VELP SCIENTIFIC
Blocks for PCR
Supplier:  VELP SCIENTIFIC
New Product
Certificates
Blocks for PCR
3 Options Available Below
Product Details & Documents
This block is ideal for large-scale PCR and qPCR workflows, including DNA/RNA amplification, sequencing preparation, genotyping, and other nucleic acid–based studies.
Engineered for excellent tube-to-wall contact, it delivers rapid heat transfer and uniform temperatures.
Compatible with ECODryBlock and external Pt100/Pt1000 probes, it allows precise monitoring and control of reaction conditions.
Product Family Options
Product Information
  • Well size
    For
    No. of Holes
    Depth
    Availability
    Price
  • VELPA00000523
    PCR-W64-V002-A block
    PCR-W64-V002-A block
    Ø 6,4 mm
    0,2 ml individual PCR tubes
    64
    20,2 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 136,00 €
    136,00 €
    1 * 1 UN
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000524
    PCR-W80-V002-A block
    PCR-W80-V002-A block
    Ø 6,6 mm
    0,2 ml PCR tube strips
    80
    15,5 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 136,00 €
    136,00 €
    1 * 1 UN
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000525
    PCR-W96-V002-A block
    PCR-W96-V002-A block
    Ø 6,6 mm
    96-well PCR plate (0,2 ml wells)
    96
    15,5 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 136,00 €
    136,00 €
    1 * 1 UN
       136,00 €
    Specifications:
    New Product